Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336618.1 | NP_001323430.1 | 9 - 10 | 2379 | 110 | |
NM_129257.4 | NP_181238.4 | 9 - 10 | 2379 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |