Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129364.4 | NP_565881.1 | 4 - 5 | 916 | 66 | |
GenBank mRNA | AY052669.1 | - | 4 - 5 | 636 | 66 | |
AY063730.1 | - | 4 - 5 | 620 | 66 | ||
AY065187.1 | - | 4 - 5 | 702 | 66 | ||
AY114636.1 | - | 4 - 5 | 620 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |