Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129404.4 | NP_181381.2 | 2 - 3 | 689 | 129 | |
GenBank mRNA | AK226301.1 | - | 2 - 3 | 466 | 129 | |
BT033030.1 | - | 2 - 3 | 448 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |