Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129611.5 | NP_181581.1 | 5 - 6 | 831 | 90 | |
GenBank mRNA | AY142634.1 | - | 5 - 6 | 675 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |