Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084563.2 | NP_001078032.1 | 2 - 3 | 988 | 102 | |
NM_001336844.1 | NP_001318392.1 | 3 - 4 | 477 | 102 | ||
NM_001336845.1 | NP_001324081.1 | 3 - 4 | 917 | 102 | ||
NM_001336846.1 | NP_001324082.1 | 3 - 4 | 894 | 102 | ||
NM_129616.4 | NP_565936.1 | 3 - 4 | 653 | 102 | ||
GenBank mRNA | BT004049.1 | - | 3 - 4 | 445 | 102 | |
BT005048.1 | - | 2 - 3 | 275 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |