Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMBP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129666.5 | NP_181634.1 | 1 | 1094 | 149 | |
GenBank mRNA | AK117111.1 | - | 1 | 851 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |