Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT-P4H-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337014.1 | NP_001324186.1 | 2 - 3 | 322 | 81 | |
NM_129869.5 | NP_181836.1 | 2 - 3 | 415 | 81 | ||
GenBank mRNA | AK117688.1 | - | 2 - 3 | 192 | 81 | |
BT006098.1 | - | 2 - 3 | 108 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |