Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084585.2 | NP_001078054.1 | 3 - 4 | 345 | 86 | |
NM_129973.3 | NP_181938.1 | 3 - 4 | 568 | 86 | ||
GenBank mRNA | AF361801.1 | - | 3 - 4 | 335 | 86 | |
AK226690.1 | - | 3 - 4 | 338 | 86 | ||
AK227643.1 | - | 3 - 4 | 335 | 86 | ||
AK316933.1 | - | 3 - 4 | 339 | 86 | ||
AY050418.2 | - | 3 - 4 | 336 | 86 | ||
AY113036.1 | - | 3 - 4 | 233 | 86 | ||
D83531.1 | - | 3 - 4 | 336 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |