Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129982.3 | NP_181947.1 | 4 - 5 | 1464 | 81 | |
GenBank mRNA | BT022041.1 | - | 4 - 5 | 1442 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |