Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129118.5 | NP_565812.1 | 2 - 3 | 507 | 173 | |
GenBank mRNA | AK118678.1 | - | 2 - 3 | 404 | 173 | |
AY074850.1 | - | 2 - 3 | 404 | 173 | ||
AY093802.1 | - | 2 - 3 | 354 | 173 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |