Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035754.1 | NP_001030831.1 | 2 - 3 | 189 | 148 | |
NM_001084791.1 | NP_001078260.1 | 2 - 3 | 164 | 148 | ||
NM_001084792.1 | NP_001078261.1 | 2 - 3 | 162 | 148 | ||
NM_001339393.1 | NP_001327179.1 | 2 - 3 | 500 | 148 | ||
NM_001339394.1 | NP_001327180.1 | 2 - 3 | 526 | 148 | ||
NM_114760.4 | NP_190470.1 | 2 - 3 | 174 | 148 | ||
NM_180341.3 | NP_850672.1 | 2 - 3 | 172 | 148 | ||
GenBank mRNA | AF428384.1 | - | 2 - 3 | 150 | 148 | |
AK317079.1 | - | 2 - 3 | 190 | 148 | ||
AY050854.1 | - | 2 - 3 | 154 | 148 | ||
AY065137.1 | - | 2 - 3 | 160 | 148 | ||
AY079381.1 | - | 1 - 2 | 106 | 148 | ||
AY081595.1 | - | 1 - 2 | 106 | 148 | ||
X75162.1 | - | 129 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |