Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339262.1 | NP_001327017.1 | 4 - 5 | 1211 | 128 | |
NM_114519.3 | NP_190236.1 | 4 - 5 | 1211 | 128 | ||
GenBank mRNA | BT003847.1 | - | 4 - 5 | 1159 | 128 | |
BT005073.1 | - | 3 - 4 | 1069 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |