Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGXT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340334.1 | NP_001328015.1 | 2 - 3 | 744 | 87 | |
NM_116407.4 | NP_192086.1 | 2 - 3 | 744 | 87 | ||
GenBank mRNA | AY924810.1 | - | 2 - 3 | 653 | 87 | |
KJ138771.1 | - | 2 - 3 | 653 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |