Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036550.1 | NP_001031627.1 | 2 - 3 | 660 | 108 | |
NM_001036551.1 | NP_001031628.1 | 2 - 3 | 645 | 108 | ||
NM_001084918.1 | NP_001078387.1 | 1 - 2 | 780 | 108 | ||
NM_117468.3 | NP_193130.1 | 1 - 2 | 968 | 108 | ||
GenBank mRNA | AF059581.1 | - | 1 - 2 | 756 | 108 | |
AF325037.2 | - | 1 - 2 | 776 | 108 | ||
AY042866.1 | - | 1 - 2 | 778 | 108 | ||
AY049279.1 | - | 1 - 2 | 776 | 108 | ||
AY081468.1 | - | 1 - 2 | 710 | 108 | ||
AY090284.1 | - | 1 - 2 | 710 | 108 | ||
BT002404.1 | - | 1 - 2 | 776 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |