Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABA1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117996.6 | NP_193615.1 | 1 - 2 | 461 | 115 | |
GenBank mRNA | AK228509.1 | - | 1 - 2 | 439 | 115 | |
BT003901.1 | - | 1 - 2 | 439 | 115 | ||
BT004936.1 | - | 1 - 2 | 227 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |