Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCP4;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120827.3 | NP_196362.1 | 1 - 2 | 433 | 121 | |
GenBank mRNA | AK119113.1 | - | 1 - 2 | 430 | 121 | |
BT005235.1 | - | 1 - 2 | 375 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |