Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GID1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122614.4 | NP_198084.1 | 2 | 1556 | 77 | |
GenBank mRNA | AY054653.1 | - | 2 | 1431 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |