Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106161.3 | NP_177641.1 | 2 | 895 | 154 | |
GenBank mRNA | L78079.1 | - | 2 | 842 | 154 | |
M90510.1 | - | 2 | 854 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |