Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106504.4 | NP_177978.1 | 2 | 600 | 101 | |
GenBank mRNA | AY042833.1 | - | 2 | 444 | 101 | |
AY081471.1 | - | 1 | 269 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |