Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036586.3 | NP_001031663.1 | 1 - 2 | 332 | 101 | |
GenBank mRNA | AK227637.1 | - | 1 - 2 | 183 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |