Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCR46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036962.2 | NP_001032039.1 | 1 - 2 | 170 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |