Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105276.4 | NP_176779.2 | 9 | 1995 | 164 | |
GenBank mRNA | AK175694.1 | - | 9 | 1851 | 164 | |
AK175887.1 | - | 9 | 1851 | 164 | ||
AK176012.1 | - | 9 | 1851 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |