Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075227.1 | NP_001068695.1 | 2 - 3 | 508 | 78 | |
GenBank mRNA | BC122712.1 | - | 2 - 3 | 508 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |