Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101044.2 | NP_001094514.2 | 5 - 6 | 927 | 85 | |
NM_001166298.1 | NP_001159770.1 | 5 - 6 | 927 | 85 | ||
GenBank mRNA | AJ299408.1 | - | 5 - 6 | 632 | 85 | |
BC149435.1 | - | 5 - 6 | 913 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |