Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001157.1 | NP_001001157.1 | 1 - 2 | 367 | 77 | |
GenBank mRNA | AB119637.1 | - | 1 - 2 | 367 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Bos taurus Probe spans exons |