Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001163.1 | NP_001001163.1 | 2 - 3 | 240 | 120 | |
GenBank mRNA | AJ699421.1 | - | 2 - 3 | 240 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |