Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPTIN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001168.1 | NP_001001168.1 | 5 - 6 | 403 | 88 | |
GenBank mRNA | AY568576.1 | - | 5 - 6 | 403 | 88 | |
BC148893.1 | - | 5 - 6 | 370 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |