Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015616.1 | NP_001015616.1 | 4 - 5 | 657 | 80 | |
GenBank mRNA | BC134590.1 | - | 4 - 5 | 679 | 80 | |
BT020717.1 | - | 4 - 5 | 657 | 80 | ||
BT020773.1 | - | 4 - 5 | 658 | 80 | ||
BT020778.1 | - | 4 - 5 | 657 | 80 | ||
BT021642.1 | - | 4 - 5 | 657 | 80 | ||
BT021761.1 | - | 4 - 5 | 657 | 80 | ||
BT025340.1 | - | 4 - 5 | 643 | 80 | ||
BT025483.1 | - | 4 - 5 | 649 | 80 | ||
BT026244.1 | - | 4 - 5 | 668 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |