Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM76A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015660.1 | NP_001015660.1 | 3 - 4 | 249 | 87 | |
GenBank mRNA | BT020680.1 | - | 3 - 4 | 249 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |