Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPZL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015673.2 | NP_001015673.1 | 5 - 6 | 902 | 144 | |
GenBank mRNA | BC102486.1 | - | 5 - 6 | 794 | 144 | |
BT020659.1 | - | 5 - 6 | 903 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |