Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017933.1 | NP_001017933.1 | 4 - 5 | 669 | 68 | |
GenBank mRNA | BC122698.1 | - | 4 - 5 | 612 | 68 | |
BT020635.1 | - | 4 - 5 | 669 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |