Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017940.1 | NP_001017940.1 | 4 - 5 | 474 | 113 | |
GenBank mRNA | AJ865078.1 | - | 4 - 5 | 474 | 113 | |
BC149487.1 | - | 6 - 7 | 767 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |