Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024511.1 | NP_001019682.1 | 9 - 10 | 1314 | 103 | |
GenBank mRNA | BC110248.1 | - | 9 - 10 | 1340 | 103 | |
BT020627.1 | - | 9 - 10 | 1314 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |