Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034034.2 | NP_001029206.1 | 10 - 11 | 904 | 68 | |
GenBank mRNA | AF077815.1 | - | 2 - 3 | 163 | 68 | |
BC102589.1 | - | 10 - 11 | 905 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |