Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173941.2 | NP_776366.1 | 14 - 15 | 2165 | 115 | |
GenBank mRNA | AF355147.1 | - | 14 - 15 | 2165 | 115 | |
BC149724.1 | - | 14 - 15 | 2179 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |