Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173942.2 | NP_776367.1 | 7 - 8 | 793 | 130 | |
GenBank mRNA | U96448.1 | - | 7 - 8 | 793 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |