Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174002.3 | NP_776427.1 | 3 - 4 | 1008 | 104 | |
GenBank mRNA | S67307.1 | - | 3 - 4 | 1012 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Bos taurus Probe spans exons |