Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174048.2 | NP_776473.1 | 5 - 6 | 496 | 106 | |
GenBank mRNA | BC149525.1 | - | 5 - 6 | 513 | 106 | |
M80838.1 | - | 5 - 6 | 501 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |