Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174137.2 | NP_776562.1 | 6 - 7 | 1113 | 57 | |
GenBank mRNA | BC103451.1 | - | 6 - 7 | 1137 | 57 | |
BT025406.1 | - | 6 - 7 | 1111 | 57 | ||
KR006346.1 | - | 5 - 6 | 992 | 57 | ||
X16383.1 | - | 1113 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |