Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174279.2 | NP_776704.1 | 1 - 2 | 242 | 81 | |
GenBank mRNA | X76485.1 | - | 242 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |