Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174290.2 | NP_776715.1 | 2 - 3 | 321 | 88 | |
GenBank mRNA | AF029793.2 | - | 2 - 3 | 321 | 88 | |
BC102745.1 | - | 2 - 3 | 368 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |