Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174312.2 | NP_776737.1 | 1 - 2 | 155 | 65 | |
GenBank mRNA | AF030029.1 | - | 1 - 2 | 155 | 65 | |
BC151475.1 | - | 1 - 2 | 130 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |