Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174324.2 | NP_776749.1 | 5 - 6 | 712 | 90 | |
GenBank mRNA | BC105419.1 | - | 6 - 7 | 754 | 90 | |
M14207.1 | - | 2 - 3 | 281 | 90 | ||
X03642.1 | - | 712 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |