Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174388.2 | NP_776813.1 | 2 - 3 | 232 | 70 | |
GenBank mRNA | M59851.1 | - | 2 - 3 | 150 | 70 | |
S68064.1 | - | 2 - 3 | 232 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |