Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174455.3 | NP_776880.1 | 2 - 3 | 114 | 97 | |
GenBank mRNA | BC102108.1 | - | 2 - 3 | 113 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |