Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174508.2 | NP_776933.1 | 1 - 2 | 135 | 131 | |
GenBank mRNA | M35551.1 | - | 1 - 2 | 135 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |