Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174620.2 | NP_777045.1 | 1 | 219 | 77 | |
GenBank mRNA | AF067370.1 | - | 1 | 219 | 77 | |
BC108160.1 | - | 3 - 4 | 240 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |