Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174628.1 | NP_777053.1 | 1 - 2 | 180 | 98 | |
GenBank mRNA | AY035552.1 | - | 1 - 2 | 180 | 98 | |
BC120031.1 | - | 1 - 2 | 404 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |