Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174667.3 | NP_777092.1 | 1 - 2 | 119 | 73 | |
GenBank mRNA | AY911318.1 | - | 1 - 2 | 99 | 73 | |
BC103425.1 | - | 1 | 99 | 73 | ||
BC151404.1 | - | 1 | 100 | 73 | ||
X63209.1 | - | 120 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |