Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174719.2 | NP_777144.1 | 5 - 6 | 1724 | 118 | |
GenBank mRNA | BC142063.1 | - | 5 - 6 | 1401 | 118 | |
X57032.1 | - | 1724 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |